siRNA Details
Gene Symbol: Nbeal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173444.2 | NP_775620.2 | 46 | 7220 | ||
GenBank mRNA | AK041108.1 | - | 6 | 558 | ||
BC055813.1 | - | 9 | 949 | |||
BC060141.1 | - | 22 | 3351 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
