siRNA Details
Gene Symbol: Slc45a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173774.3 | NP_776135.2 | 5 | 1339 | ||
GenBank mRNA | AK082651.1 | - | 1332 | |||
AK134677.1 | - | 5 | 1265 | |||
AK134683.1 | - | 5 | 1330 | |||
AK158002.1 | - | 5 | 1329 | |||
BC051188.1 | - | 5 | 1301 |
