siRNA Details
Gene Symbol: Ticrr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029835.1 | NP_084111.1 | 20 | 5026 | ||
GenBank mRNA | AK033104.1 | - | 1 | 572 | ||
BC058634.1 | - | 1 | 238 | |||
BC158103.1 | - | 20 | 4990 | 9088180 |

More Information
Other Species Targeted

Gene Ontology Categories:
