siRNA Details
Gene Symbol: Ticrr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029835.1 | NP_084111.1 | 22 | 6430 | ||
GenBank mRNA | AK137597.1 | - | 21 | 4313 | ||
BC028450.1 | - | 1 | 636 | |||
BC058634.1 | - | 3 | 1642 | |||
BC158103.1 | - | 22 | 6394 | 9088180 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
