siRNA Details
Gene Symbol: St18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244692.1 | NP_001231621.1 | 24 | 4449 | ||
NM_001244693.1 | NP_001231622.1 | 23 | 4371 | |||
NM_173868.2 | NP_776293.1 | 22 | 4262 | |||
GenBank mRNA | AJ536155.1 | - | 24 | 4413 | ||
AK122303.1 | - | 22 | 4211 | |||
AK147320.1 | - | 24 | 4450 | |||
AK147664.1 | - | 24 | 4450 | |||
BC032273.1 | - | 17 | 3176 |
