siRNA Details
Gene Symbol: Adam6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174885.3 | NP_777479.2 | 1 | 422 | ||
GenBank mRNA | AY158689.1 | - | 1 | 422 | ||
BC108387.1 | - | 1 | 375 | |||
BC139059.1 | - | 1 | 407 | 8862081 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
