siRNA Details
Gene Symbol: Zfp804a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175513.3 | NP_780722.2 | 4 | 1991 | ||
GenBank mRNA | AK035949.1 | - | 1 | 513 | ||
BC035535.1 | - | 1 | 1018 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
