siRNA Details
Gene Symbol: C130060K24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175524.4 | NP_780733.2 | 6 | 2529 | ||
GenBank mRNA | AK048439.1 | - | 6 | 2439 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C130060K24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175524.4 | NP_780733.2 | 6 | 2529 | ||
GenBank mRNA | AK048439.1 | - | 6 | 2439 |