siRNA Details
Gene Symbol: Scaper | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081341.1 | NP_001074810.1 | 25 | 3115 | ||
GenBank mRNA | AK048755.1 | - | 14 | 1643 | ||
AK173175.1 | - | 24 | 3045 | |||
BC157959.1 | - | 24 | 3060 | 9007286 | ||
BC158062.1 | - | 24 | 3037 | 9088165 | ||
BC172119.1 | - | 24 | 3060 | 9054405 | ||
BC172151.1 | - | 24 | 3037 | 9054821 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
