siRNA Details
Gene Symbol: Scaper | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081341.1 | NP_001074810.1 | 27 | 3324 | ||
GenBank mRNA | AK048755.1 | - | 16 | 1852 | ||
AK173175.1 | - | 26 | 3254 | |||
BC052188.1 | - | 1 | 64 | |||
BC157959.1 | - | 26 | 3269 | 9007286 | ||
BC158062.1 | - | 26 | 3246 | 9088165 | ||
BC172119.1 | - | 26 | 3269 | 9054405 | ||
BC172151.1 | - | 26 | 3246 | 9054821 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
