siRNA Details
Gene Symbol: Nlrc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081280.1 | NP_001074749.1 | 18 | 3741 | ||
NM_175547.3 | NP_780756.1 | 15 | 1664 | |||
GenBank mRNA | AK084199.1 | - | 15 | 1578 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
