siRNA Details
Gene Symbol: Nlrc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081280.1 | NP_001074749.1 | 13, 14 | 3139 | ||
NM_175547.3 | NP_780756.1 | 10, 11 | 1062 | |||
GenBank mRNA | AK079766.1 | - | 10, 11 | 986 | ||
AK084199.1 | - | 10, 11 | 976 | |||
AK171971.1 | - | 12, 13 | 4152 | |||
BC130223.1 | - | 10, 11 | 2528 | |||
BC152543.1 | - | 10, 11 | 2537 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
