siRNA Details
Gene Symbol: Dido1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175551.4 | NP_780760.2 | 16 | 5426 | ||
GenBank mRNA | AK031764.1 | - | 1 | 464 | ||
AK032843.1 | - | 1 | 649 | |||
AY425952.1 | - | 16 | 5328 | |||
BC039702.1 | - | 1 | 30 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
