siRNA Details
Gene Symbol: Nfib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113209.2 | NP_001106680.1 | 12 | 8060 | ||
NM_001113210.2 | NP_001106681.1 | 12 | 8033 | |||
NM_001286127.1 | NP_001273056.1 | 11 | 7944 | |||
NM_001286131.1 | NP_001273060.1 | 10 | 7811 | |||
NM_008687.6 | NP_032713.3 | 9 | 7722 | |||
GenBank mRNA | AK030686.1 | - | 1 | 1268 | ||
AK050276.1 | - | 1 | 2305 | |||
AK053181.1 | - | 1 | 4109 | |||
AK081555.1 | - | 1 | 4110 | |||
BC031188.1 | - | 1 | 1070 | |||
BC096542.1 | - | 1 | 1427 |

More Information
Other Species Targeted

Gene Ontology Categories:
