siRNA Details
Gene Symbol: 9130019O22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030226.3 | NP_084502.2 | 3 | 779 | ||
GenBank mRNA | AK020267.1 | - | 3 | 654 | ||
AK035847.1 | - | 3 | 781 | |||
BC031466.1 | - | 3 | 749 | 4038184 |
Gene Symbol: Zfp747 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175560.3 | NP_780769.1 | 3 | 794 | ||
GenBank mRNA | AK032584.1 | - | 3 | 760 | ||
AK042972.1 | - | 3 | 795 | |||
BC058799.1 | - | 3 | 740 | 6484930 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
