siRNA Details
Gene Symbol: Pcnxl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175561.4 | NP_780770.2 | 19, 20 | 3691 | ||
GenBank mRNA | AK087907.1 | - | 4, 5 | 604 | ||
AK220342.1 | - | 11, 12 | 1253 | |||
BC068235.1 | - | 8, 9 | 1760 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
