siRNA Details
Gene Symbol: Pcnxl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175561.4 | NP_780770.2 | 26, 27 | 4794 | ||
GenBank mRNA | AK030215.1 | - | 1, 2 | 338 | ||
AK087907.1 | - | 11, 12 | 1710 | |||
AK220342.1 | - | 18, 19 | 2356 | |||
BC068235.1 | - | 15, 16 | 2861 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
