siRNA Details
Gene Symbol: Zfp738 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001187.3 | NP_001001187.2 | 4 | 3430 | ||
GenBank mRNA | AK032995.1 | - | 1 | 1970 | ||
BC082605.1 | - | 4 | 3385 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
