siRNA Details
Gene Symbol: Zfp608 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175751.4 | NP_786927.2 | 5, 6 | 4240 | ||
GenBank mRNA | AK076178.1 | - | 1, 2 | 1116 | ||
AK077030.1 | - | 4, 5 | 2967 | |||
BC059268.1 | - | 4, 5 | 3907 | |||
BC092219.1 | - | 5, 6 | 3973 | 6830664 | ||
BC151073.1 | - | 4, 5 | 3759 | 9087986 | ||
BC151075.1 | - | 4, 5 | 3759 | 9087988 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
