siRNA Details
Gene Symbol: Nwd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176940.5 | NP_795914.3 | 5 | 1101 | ||
GenBank mRNA | AK034868.1 | - | 800 | |||
AK038792.1 | - | 4 | 793 | |||
AK087622.1 | - | 5 | 1103 | |||
AK162888.1 | - | 5 | 1096 | |||
BC082552.1 | - | 5 | 1041 | |||
BC144786.1 | - | 3 | 691 | 9053334 | ||
BC144787.1 | - | 3 | 691 | |||
BC145703.1 | - | 3 | 691 | 40131006 |
