siRNA Details
Gene Symbol: Syne2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005510.2 | NP_001005510.2 | 10 | 1184 | ||
GenBank mRNA | AK052521.1 | - | 10 | 1153 | ||
AK078542.1 | - | 10 | 1185 | |||
AK084394.1 | - | 11 | 1364 | |||
AK145252.1 | - | 10 | 1185 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
