siRNA Details
Gene Symbol: Uggt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081252.2 | NP_001074721.2 | 28 | 3315 | ||
GenBank mRNA | AK046834.1 | - | 16 | 1990 | ||
BC110672.1 | - | 14 | 1696 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
