siRNA Details
Gene Symbol: 9330133O14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK033985.1 | - | 1 | 1466 | ||
AK049744.1 | - | 1 | 1524 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_176948.1 | - | 1524 | Chr. - - [-] on Build |
