siRNA Details
Gene Symbol: Simc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176987.4 | NP_795961.3 | 5, 6 | 3355 | ||
GenBank mRNA | AK028926.1 | - | 5, 6 | 1020 | ||
AK052561.1 | - | 4, 5 | 1149 | |||
AK166176.1 | - | 4, 5 | 959 | |||
BC059035.1 | - | 4, 5 | 1614 | |||
BC094550.1 | - | 4, 5 | 1166 |
