siRNA Details
Gene Symbol: Simc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176987.4 | NP_795961.3 | 10 | 4647 | ||
GenBank mRNA | AK028926.1 | - | 10 | 2312 | ||
BC023054.1 | - | 5 | 1267 | |||
BC059035.1 | - | 9 | 2906 | |||
BC094550.1 | - | 9 | 2458 |
