siRNA Details
Gene Symbol: Atp10b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176999.3 | NP_795973.2 | 2 | 316 | ||
GenBank mRNA | AK033531.1 | - | 2 | 316 |

More Information
Other Species Targeted

Gene Ontology Categories:
