siRNA Details
Gene Symbol: A530016L24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177039.4 | NP_796013.1 | 2, 3 | 392 | ||
GenBank mRNA | AK040701.1 | - | 2, 3 | 372 | ||
AK040838.1 | - | 2, 3 | 369 | |||
AK079947.1 | - | 2, 3 | 374 | |||
AK163065.1 | - | 2, 3 | 394 |
