siRNA Details
Gene Symbol: A530016L24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177039.4 | NP_796013.1 | 5 | 1088 | ||
GenBank mRNA | AK040701.1 | - | 5 | 1068 | ||
AK040838.1 | - | 4 | 1539 | |||
AK079947.1 | - | 5 | 1071 | |||
AK163065.1 | - | 5 | 1090 | |||
BC026527.1 | - | 5 | 921 | 4924828 | ||
BC096642.1 | - | 5 | 1057 | 1067695 |
