siRNA Details
Gene Symbol: Auts2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177047.3 | NP_796021.2 | 1 | 775 | ||
GenBank mRNA | AK052418.1 | - | 1 | 837 | ||
AK077988.1 | - | 1 | 1232 | |||
AK085946.1 | - | 1 | 464 | |||
AK147478.1 | - | 1774 | ||||
BC096021.1 | - | 1 | 774 | |||
BC151031.1 | - | 1 | 789 | |||
BC151047.1 | - | 1 | 788 |

More Information
Other Species Targeted

Gene Ontology Categories:
