siRNA Details
Gene Symbol: Jph4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177049.5 | NP_796023.2 | 6 | 3434 | ||
GenBank mRNA | AB096263.1 | - | 4 | 1877 | ||
AB099712.1 | - | 6 | 2777 | |||
AK048607.1 | - | 5 | 2535 | |||
AK148135.1 | - | 5 | 2605 | |||
AY341261.1 | - | 6 | 3434 | |||
BC052049.1 | - | 5 | 2479 | 5689347 | ||
BC052376.1 | - | 6 | 2531 | 5696231 | ||
BC052722.1 | - | 6 | 2505 | 6810400 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
