siRNA Details
Gene Symbol: Jph4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177049.5 | NP_796023.2 | 6 | 3550 | ||
GenBank mRNA | AB096263.1 | - | 4 | 1993 | ||
AB099712.1 | - | 6 | 2893 | |||
AK048607.1 | - | 5 | 2651 | |||
AK148135.1 | - | 5 | 2721 | |||
AY341261.1 | - | 6 | 3550 | |||
BC052049.1 | - | 5 | 2595 | 5689347 | ||
BC052376.1 | - | 6 | 2647 | 5696231 | ||
BC052722.1 | - | 6 | 2621 | 6810400 |

More Information
Other Species Targeted

Gene Ontology Categories:
