siRNA Details
Gene Symbol: A630091E08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK080303.1 | - | 2 | 730 | ||
AK080393.1 | - | 2 | 732 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: A630091E08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK080303.1 | - | 2 | 730 | ||
AK080393.1 | - | 2 | 732 |