siRNA Details
Gene Symbol: SPTLC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004863.3 | NP_004854.1 | 2 | 353 | ||
XM_011537384.2 | XP_011535686.1 | 2 | 249 | |||
GenBank mRNA | AB011098.1 | - | 2 | 353 | ||
BC005123.2 | - | 2 | 254 | 3688136 | ||
Y08686.1 | - | 2 | 213 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
