siRNA Details
Gene Symbol: Kalrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177357.3 | NP_796331.2 | 60 | 8688 | ||
GenBank mRNA | AK081504.1 | - | 2 | 1457 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
