siRNA Details
Gene Symbol: AI854703 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK038960.1 | - | 2 | 1396 | ||
AK039299.1 | - | 1109 | ||||
AK129002.1 | - | 2 | 777 | |||
BC145738.1 | - | 2 | 1160 | |||
BC145740.1 | - | 2 | 1160 |
