siRNA Details
Gene Symbol: BC048671 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177738.2 | NP_808406.1 | 5 | 867 | ||
GenBank mRNA | AK077180.1 | - | 5 | 869 | ||
BC048671.1 | - | 4 | 581 | 6774036 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: BC048671 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177738.2 | NP_808406.1 | 5 | 867 | ||
GenBank mRNA | AK077180.1 | - | 5 | 869 | ||
BC048671.1 | - | 4 | 581 | 6774036 |