siRNA Details
Gene Symbol: BC035947 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK032005.1 | - | 2 | 881 | ||
BC034187.1 | - | 3 | 821 | 4975064 | ||
BC035948.1 | - | 1 | 118 | |||
BC138135.1 | - | 2 | 835 | 8861154 | ||
BC138136.1 | - | 2 | 835 | 8861155 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
