siRNA Details
Gene Symbol: Rnf214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178709.4 | NP_848824.1 | 4 | 1175 | ||
GenBank mRNA | AK028768.1 | - | 4 | 810 | ||
AK030208.1 | - | 4 | 807 | |||
AK032376.1 | - | 4 | 241 | |||
AK033867.1 | - | 4 | 671 | |||
AK046190.1 | - | 4 | 1160 | |||
AK046997.1 | - | 800 | ||||
AK051527.1 | - | 4 | 1176 | |||
BC049909.1 | - | 4 | 316 | |||
BC094247.1 | - | 3 | 660 | 5717984 |
