siRNA Details
Gene Symbol: Rnf214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178709.4 | NP_848824.1 | 9, 10 | 1780 | ||
GenBank mRNA | AK028768.1 | - | 9, 10 | 1415 | ||
AK030208.1 | - | 9, 10 | 1412 | |||
AK032376.1 | - | 9, 10 | 846 | |||
AK033867.1 | - | 9, 10 | 1276 | |||
AK051527.1 | - | 9, 10 | 1781 | |||
AK078706.1 | - | 4, 5 | 314 | |||
AK136229.1 | - | 4, 5 | 305 | |||
BC023073.1 | - | 1, 2 | 58 | |||
BC049909.1 | - | 9, 10 | 921 | |||
BC094247.1 | - | 8, 9 | 1265 | 5717984 |
