siRNA Details
Gene Symbol: Phf20l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081409.1 | NP_001074878.1 | 19 | 3000 | ||
GenBank mRNA | AK046670.1 | - | 2 | 520 | ||
AK053606.1 | - | 9 | 1359 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
