siRNA Details
Gene Symbol: Phf20l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081409.1 | NP_001074878.1 | 21 | 3982 | ||
GenBank mRNA | AK046670.1 | - | 4 | 1502 | ||
AK053606.1 | - | 11 | 2341 |

More Information
Other Species Targeted

Gene Ontology Categories:
