siRNA Details
Gene Symbol: MAPK11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002751.6 | NP_002742.3 | 9 | 844 | ||
NR_110887.1 | 9 | 844 | ||||
XM_011530706.1 | XP_011529008.1 | 7 | 568 | |||
GenBank mRNA | AF001008.1 | - | 9 | 743 | ||
AF001174.1 | - | 9 | 749 | |||
AF031135.1 | - | 9 | 758 | |||
AK291845.1 | - | 9 | 795 | |||
AK299745.1 | - | 6 | 1312 | |||
AK308969.1 | - | 9 | 760 | |||
AK309046.1 | - | 7 | 1502 | |||
BC027933.1 | - | 9 | 801 | 5199628 | ||
CR456514.1 | - | 9 | 745 | |||
U53442.1 | - | 9 | 786 | |||
U92268.1 | - | 9 | 747 | |||
Y14440.1 | - | 9 | 749 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
