siRNA Details
Gene Symbol: CREB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004904.3 | NP_004895.2 | 3 | 169 | ||
NM_182898.3 | NP_878901.2 | 3 | 488 | |||
NM_182899.4 | NP_878902.2 | 2 | 144 | |||
XM_005249906.1 | XP_005249963.1 | 3 | 164 | |||
XM_017012806.1 | XP_016868295.1 | 4 | 219 | |||
XM_017012807.1 | XP_016868296.1 | 4 | 1341 | |||
XM_017012808.1 | XP_016868297.1 | 4 | 200 | |||
XM_017012809.1 | XP_016868298.1 | 4 | 225 | |||
GenBank mRNA | AB451409.1 | - | 2 | 98 | ||
AK292916.1 | - | 2 | 157 | |||
BG199550.1 | - | 95 | ||||
BX282747.1 | - | 4 | 206 | |||
L05515.1 | - | 3 | 488 | |||
L05911.1 | - | 3 | 170 | |||
L05913.1 | - | 2 | 144 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: AK035031, AK051275, AK084024, AK166503, AK034642, and others.
Gene Ontology Categories:
