siRNA Details
Gene Symbol: Spin4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178753.4 | NP_848868.1 | 1 | 1337 | ||
GenBank mRNA | AK033181.1 | - | 1 | 654 | ||
AK036158.1 | - | 1 | 1338 | |||
AK043030.1 | - | 1 | 1305 | |||
AK047896.1 | - | 1 | 1298 | |||
AK147852.1 | - | 1 | 1304 | |||
AK155170.1 | - | 1 | 1277 | |||
BC027796.1 | - | 1 | 1303 | 5323183 |
