siRNA Details
Gene Symbol: Zfp750 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178763.4 | NP_848878.1 | 2 | 1319 | ||
GenBank mRNA | AK028879.1 | - | 2 | 1268 | ||
AK030690.1 | - | 2 | 1274 | |||
AK037185.1 | - | 2 | 1280 | |||
AK037240.1 | - | 2 | 1265 | |||
AK076268.1 | - | 2 | 1313 | |||
AK140792.1 | - | 1312 | ||||
BC080834.1 | - | 2 | 1290 | |||
BC150905.1 | - | 1 | 1173 | 9087816 |
