siRNA Details
Gene Symbol: Sbf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177324.2 | NP_796298.2 | 14 | 1662 | ||
GenBank mRNA | AK046622.1 | - | 14 | 1650 | ||
AK048952.1 | - | 14 | 1639 | |||
AK080952.1 | - | 14 | 1663 | |||
AK080963.1 | - | 14 | 1657 | |||
AK087396.1 | - | 14 | 1657 | |||
AK173257.1 | - | 14 | 1621 | |||
BC067204.1 | - | 15 | 1700 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
