siRNA Details
Gene Symbol: ABCG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004915.3 | NP_004906.3 | 2 | 329 | ||
NM_016818.2 | NP_058198.2 | 2 | 329 | |||
NM_207174.1 | NP_997057.1 | 2 | 330 | |||
NM_207627.1 | NP_997510.1 | 3 | 489 | |||
NM_207628.1 | NP_997511.1 | 4 | 407 | |||
NM_207629.1 | NP_997512.1 | 2 | 293 | |||
XM_011529806.1 | XP_011528108.1 | 2 | 407 | |||
XM_011529807.2 | XP_011528109.1 | 2 | 407 | |||
GenBank mRNA | AF323660.1 | - | 3 | 489 | ||
AF323661.1 | - | 4 | 407 | |||
AF323662.1 | - | 2 | 293 | |||
AF323663.1 | - | 2 | 252 | |||
AF323664.1 | - | 2 | 330 | |||
AK292152.1 | - | 2 | 339 | |||
AK298491.1 | - | 2 | 407 | |||
BC029158.1 | - | 2 | 329 | 5198758 | ||
X91249.1 | - | 2 | 241 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
