siRNA Details
Gene Symbol: Thada | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183021.3 | NP_898842.2 | 30 | 4411 | ||
GenBank mRNA | AK155218.1 | - | 1 | 87 | ||
AK170056.1 | - | 1 | 103 | |||
AK173258.1 | - | 15 | 1921 | |||
BC052885.1 | - | 5 | 551 | |||
EF222207.1 | - | 30 | 4319 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
