siRNA Details
Gene Symbol: Thada | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183021.3 | NP_898842.2 | 38 | 6063 | ||
GenBank mRNA | AK078353.1 | - | 7 | 1466 | ||
AK155218.1 | - | 9 | 1739 | |||
AK170056.1 | - | 9 | 1756 | |||
BC052885.1 | - | 13 | 2203 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
