siRNA Details
Gene Symbol: Nup54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183392.2 | NP_899248.1 | 3 | 402 | ||
GenBank mRNA | AK014260.1 | - | 3 | 295 | ||
AK087827.1 | - | 3 | 296 | |||
AK088855.1 | - | 3 | 327 | |||
AK133841.1 | - | 3 | 400 | |||
AK150182.1 | - | 3 | 327 | |||
AK150841.1 | - | 3 | 288 | |||
AK151896.1 | - | 3 | 288 | |||
AK152136.1 | - | 3 | 288 | |||
AK162037.1 | - | 3 | 334 | |||
AK207248.1 | - | 1 | 25 | |||
BC044303.1 | - | 3 | 327 | |||
BC068114.1 | - | 3 | 287 | 5707888 | ||
CJ049542.1 | - | 3 | 405 |
